missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SV2C Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572189
This item is not returnable.
View return policy
Description
SV2C Polyclonal Antibody for Western Blot
Plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles.
Specifications
| SV2C | |
| Polyclonal | |
| Unconjugated | |
| SV2C | |
| 4930527L09Rik; AI841971; KIAA1054; SV2C; Synaptic vesicle glycoprotein 2C; synaptic vesicle protein 2C | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 22987, 29643 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q496J9, Q9Z2I6 | |
| SV2C | |
| KLH conjugated synthetic peptide between 547-580 amino acids from the C-terminal region of human SV2C. | |
| 400 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction