missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SV2C Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559290
This item is not returnable.
View return policy
Description
Immunogen sequence: GEYQGIPSMN QAKDSIVSVG QPKGDEYKDR RELESERRAD EEELAQQYE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles.
Specifications
| SV2C | |
| Polyclonal | |
| Unconjugated | |
| SV2C | |
| 4930527L09Rik; AI841971; KIAA1054; SV2C; Synaptic vesicle glycoprotein 2C; synaptic vesicle protein 2C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22987 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q496J9 | |
| SV2C | |
| Recombinant protein corresponding to Human SV2C. Recombinant protein control fragment (Product #RP-96839). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction