missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SV40 Large T Antigen Antibody (LP7N), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50312APC
This item is not returnable.
View return policy
Description
SV40 Large T Antigen Monoclonal antibody specifically detects SV40 Large T Antigen in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| SV40 Large T Antigen | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Immunohistochemistry, Immunocytochemistry | |
| LP7N | |
| Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| Large T antigen, Small T antigen | |
| Detergent insoluble fraction of SVK14 cell line (SV40 transformed neonatal keratinocytes) | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction