Learn More
SV40 Small T antigen Antibody (pab 280), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61248JF646
Product Code. 18207472
  • € 437.00 / 0.10mL
Description
SV40 Small T antigen Monoclonal specifically detects SV40 Small T antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SV40SmallTantigen
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
A purified Pab 419-SV40 Small T antigen
0.1 mL
Primary
Store at 4°C in the dark.
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
pab 280
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Mouse
Protein A purified
Cancer, Virology Bacteria and Parasites
Virus