missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SV40 T-antigen Antibody (Pab 430), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50598MFV500
This item is not returnable.
View return policy
Description
SV40 T-antigen Monoclonal antibody specifically detects SV40 T-antigen in Virus samples. It is validated for Western Blot, Immunoprecipitation, Radioimmune Assays
Specifications
| SV40 T-antigen | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| SV40 transformed mouse cells | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG3 |
| Western Blot, Immunoprecipitation, Radioimmune Assays (RIA) | |
| Pab 430 | |
| Western Blot, Immunoprecipitation, Radioimmunoassay | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction