missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SV40 T-Antigen Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112037
This item is not returnable.
View return policy
Description
SV40 T-Antigen Polyclonal Antibody for ICC/IF, Western Blot
The SV40 T antigen is encoded by the early region of the SV40 genome. The large T antigen binds DNA, and complexes with a 53,000 dalton cellular protein, p53, which is required for initiation of viral DNA replication during lytic growth. In addition, the large T antigen binds DNA polymerase and the transcription factor AP-2 and forms a specific complex with the P105 product of the retinoblastoma susceptibility gene.
Specifications
| SV40 T-Antigen | |
| Polyclonal | |
| Unconjugated | |
| simian vacuolating virus 40; simian virus; Simian virus 40; SV40 T-antigen; sv40 virus large t antigen | |
| Recombinant protein encompassing a sequence within the N-terminus region of SV40 T-antigen (SV40 virus (strain 776)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.14 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction