Learn More
SVL™ Bearing Stirrer for SVL™ Stirrer Shaft
Click to view available options
Standard Taper Joint:
19/26
29/32
30
45/40
Unit Size:
Each
Pack of 5
Description
- Borosilicate glass complete with fittings
- Glands manufactured from PTFE based polymer which is self lubricating and corrosion-resistant
- Two types available: With SVL™ screwthread to allow connection to SVL™ flasks, With Quickfit™ ground glass cone to allow connection to Quickfit™ flasks
Specifications
Specifications
| For Use With (Application) | SVL stirrer shaft |
| Includes | Quickfit ground Glass cone to allow connection to Quickfit flask |
| Standard Taper Joint | 30 |