Learn More
SVL™ Bearing Stirrer for SVL™ Stirrer Shaft
Product Code. 15557730
Click to view available options
Standard Taper Joint:
19/26
29/32
30
45/40
Unit Size:
Each
Pack of 5
Product Code. 15557730
Brand: SVL 58812
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to accept SVL™ stirrer shafts with ground glass surfaces.
  • Borosilicate glass complete with fittings
  • Glands manufactured from PTFE based polymer which is self lubricating and corrosion-resistant
  • Two types available: With SVL™ screwthread to allow connection to SVL™ flasks, With Quickfit™ ground glass cone to allow connection to Quickfit™ flasks
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SVL stirrer shaft
Includes Quickfit ground Glass cone to allow connection to Quickfit flask
Standard Taper Joint 45/40
Material PTFE Based Polymer