Learn More
Quickfit™ Claisen Still Head Adapter
Click to view available options
Joint Type:
Condenser: 19/26, Flask: 24/29
Condenser: 24/29, Flask: 24/29
Size:
Socket: 14/23
Unit Size:
Each
Pack of 5
Description
- Similar to the SH4: still heads but with extra socket to accept dropping funnel or stirrer
Specifications
Specifications
| Type | Claisen Still Head |
| Material | Borosilicate Glass |
| Joint Type | Condenser: 19/26, Flask: 24/29 |
| Size | Socket: 14/23 |