Learn More
Invitrogen™ SVOPL Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5113126
Product Code. 17170984
  • € 458.00 / 100µg
Description
SVOPL Polyclonal Antibody for Western Blot, ICC/IF, ELISA
SVOPL (putative transporter SVOPL), also known as SV2-related protein-like, is a 492 amino acid multi-pass membrane protein belonging to the major facilitator superfamily. SVOPL is a paralog to synaptic vesicle protein (SVOP) that exists as two alternatively spliced isoforms. The gene encoding SVOPL maps to human chromosome 7q34. Chromosome 7 is approximately 158 milllion bases long, encodes over 1,000 genes and makes up approximately 5% of the human genome. Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. Deletions of portions of the q arm of chromosome 7 are linked to myeloid disorders, including acute myelogenous leukemia and myelodysplasia.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SVOPL
Polyclonal
Unconjugated
SVOPL
9430071P14Rik; LOC518832 protein; putative transporter SVOPL; RGD1566053; si:dkeyp-27e10.2; similar to RIKEN cDNA 9430071P14 gene; SV2 related protein homolog (rat)-like; SV2 related protein homolog-like; SV2-related protein-like; SVOP like; Svopl; SVOP-like; SVOP-like protein; zgc:101741
Rabbit
Protein G
RUO
136306, 320590, 500085
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot, Immunocytochemistry
3 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q6PDF3, Q8N434
SVOPL
Recombinant Human Putative transporter SVOPL protein (73-129AA).