Learn More
Neogen Swab-Sampler
Product Code. p-7230034
Encompass
Click to view available options
Includes:
10 mL Neutralizing Buffer
Unit Size:
100 sets
Product Code. 17213734
Brand: Neogen 7000053818
This item requires a price update prior to being ordered.
Request a Price for this item here.
Convenient and quick collection device for environmental sampling and testing over small surface areas.
  • Ready to use for pathogen testing of environmental samples.
  • Swab reaches into corners and hard-to-reach areas for increased accuracy.
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
  • Bottles have ample headspace for sample addition and vortexing.
  • Self-contained swab and vial for increased productivity and efficiency.
Specifications
Includes 10 mL Neutralizing Buffer
No. per Case 100
Product Type Swab