missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Swine Influenza A H1N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41109
This item is not returnable.
View return policy
Description
Swine Influenza A H1N1 Hemagglutinin Polyclonal antibody specifically detects Swine Influenza A H1N1 Hemagglutinin in Virus samples. It is validated for ELISA.
Specifications
| Swine Influenza A H1N1 Hemagglutinin | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide from the novel swine influenza Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product NBP2-41107 and NBP2-41105. This antibody is a cognate pair with product number NBP2-41108. The immunogen is located within amino acids 150 - 200 of Swine H1N1 Hemagglutinin. Amino Acid Squence: VKKGNSYPK | |
| 0.1 mg | |
| Primary | |
| Virus | |
| IgG |
| ELISA | |
| 1 mg/mL | |
| ELISA 1 ug/ml | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| This antibody is specific for the novel swine influenza Hemagglutinin and will not recognize the corresponding Hemagglutinin sequence from the seasonal H1N1 influenza (A/Brisbane/59/2007 (H1N1)). Will not cross-react with peptide corresponding to the seasonal H1N1 Hemagglutinin. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction