missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Swine Influenza A H1N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41105-0.025mg
This item is not returnable.
View return policy
Description
Swine Influenza A H1N1 Hemagglutinin Polyclonal antibody specifically detects Swine Influenza A H1N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Swine Influenza A H1N1 Hemagglutinin | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide from the novel swine influenza Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product NBP2-41107 and NBP2-41109. This antibody is a cognate pair with product NBP2-41104. The immunogen is located within amino acids 120 - 170 of Swine H1N1 Hemagglutinin. Amino Acid Squence: CNHDSNKGTVAA | |
| 0.025 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction