missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Swine Influenza A H1N1 Hemagglutinin Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41105F
This item is not returnable.
View return policy
Description
Swine Influenza A H1N1 Hemagglutinin Polyclonal antibody specifically detects Swine Influenza A H1N1 Hemagglutinin in Virus samples. It is validated for ELISA
Specifications
| Swine Influenza A H1N1 Hemagglutinin | |
| Polyclonal | |
| ELISA | |
| Blue | |
| Antibody was raised against a synthetic peptide from the novel swine influenza Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product NBP2-41107 and NBP2-41109. This antibody is a cognate pair with product NBP2-41104. The immunogen is located within amino acids 120 - 170 of Swine H1N1 Hemagglutinin. Amino Acid Squence: CNHDSNKGTVAA | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Virus | |
| Purified |
| ELISA | |
| FITC | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction