missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Swine Influenza A H1N1 Nucleocapsid Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41072JF646
This item is not returnable.
View return policy
Description
Swine Influenza A H1N1 Nucleocapsid Polyclonal antibody specifically detects Swine Influenza A H1N1 Nucleocapsid in Virus samples. It is validated for ELISA, Immunofluorescence
Specifications
| Swine Influenza A H1N1 Nucleocapsid | |
| Polyclonal | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| HaCaT UV-repressible serpin, Headpin, HSHUR7SEQ, HUR7, HURPIN, MGC126870, Peptidase inhibitor 13, PI-13, PI13hurpin, protease inhibitor 13 (hurpin, headpin), Proteinase inhibitor 13, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 13, serpin B13, serpin peptidase inhibitor, clade B (ovalbumin), member 13, UV-B repressed sequence, HUR 7 | |
| Antibody was raised against a synthetic peptide from the swine-Origin H1N1 NP protein. The immunogen is located within amino acids 350 - 400 of Swine H1N1 Nucleocapsid Protein. Amino Acid Squence: AENVETMDSNT | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Virus | |
| Purified |
| ELISA, Immunofluorescence | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction