Learn More
OHAUS™ Swivel Multi-Purpose Clamps
Shop All Ohaus Products
missing translation for 'orderingAttributeHoverText'
Size (Metric) Grip:
0 to 69 mm
0 to 76 mm
0 to 95 mm
Jaw Type:
2-Prong Single Adjust
3-Prong Dual Adjust
Material:
Nickel-plated
Stainless Steel
missing translation for 'unitSize'
Each
Description
- Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
- Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
- Available as single or dual-adjust in nickel-plated zinc or stainless steel
- Non-slip vinyl sleeves, and fiberglass covers for temperatures above 100°C (included with all 2 and 3-prong clamps)
Specifications
Specifications
| Type | Multi-purpose Clamp |
| Material | Nickel-plated |
| Includes | Non-slip vinyl sleeves and fiberglass covers |
| Description | Clamp, Multi Purpose, CLM-SWIVL2SZL |
| Length (English) | 7.09 in. |
| Length (Metric) | 180 mm |
| Size | Large |
| Size (Metric) Grip | 0 to 95 mm |
| Jaw Type | 2-Prong Single Adjust |