Learn More
BOLA Swivelling Screw Fittings with Ground Joint
Product Code. p-200109930
Click to view available options
Diameter (Metric):
6 mm
10 mm
Unit Size:
Each
Product Code. 10584995
Brand: BOLA D69234
Only null left
Add to basket
Add to basket
Swivelling Screw Fittings with Ground Joint PPS cap, inner parts made of PTFE
  • Ground joint body made of PTFE, screw cap made of PPS.
  • Probes, tubes or tubing are firstly inserted through the fitting, then the fitting is put into the ground joint and the inserted element can be fixed in the requested position.
  • Suitable for all kinds of probes, tubes or hard-walled tubing
  • Can be used with GL threads or ground joints
  • Angle and height can be fixed tightly
  • A damage of tube or probe due to collision with the stirrer shaft can be avoided
Specifications
Certifications/Compliance FDA
Diameter (Metric) 6 mm
Autoclavable Autoclavable
Arm Angle
Color White
Flexibility Rigid