missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565490
This item is not returnable.
View return policy
Description
Immunogen sequence: TVAESAEEELQ QAGDQELLHQ AKDFGNYLFN FASAATKKIT ESVAETAQTI KKSVEEGKID GIIDKTIIGD FQKEQKKFVE EQHTKKSEAA VPPWVDTND Highest antigen sequence identity to the following orthologs - mouse 75%, rat 78%.
SYAP1 plays a role in adipocyte differentiation by promoting mTORC2-mediated phosphorylation of AKT1 at 'Ser-473' after growth factor stimulation.
Specifications
| SYAP1 | |
| Polyclonal | |
| Unconjugated | |
| SYAP1 | |
| 2010110O17Rik; AW011796; BSD domain-containing signal transducer and Akt interactor protein; BSTA; PRO3113; SAP47 homolog; SYAP1; synapse associated protein 1; synapse associated protein 1, SAP47 homolog; synapse-associated protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 94056 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96A49 | |
| SYAP1 | |
| Recombinant Human SYAP1. Recombinant protein control fragment (Product #RP-106167). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction