missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYCP3, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00050511-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MVSSGKKYSRKSGKPSVEDQFTRAYDFETEDKKDLSGSEEDVIEGKTAVIEKRRKKRSSAGVVEDMGGEVQNMLEGVGVDINKALLAKRKRLEMYTKASLKTSNQKIEHVWKTQQDQRQKLNQEYSQQFLTLFQQWDLDMQKAEEQEEKILNMFRQQQKILQQSRIVQSQRLKTIKQLYEQFIKSMEELEKNHDNLLTGAQNEFKKEMAMLQKKIMMETQQQEIASVRKSLQSMLFSpecifications
| SYCP3 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_153694 | |
| SYCP3 | |
| SYCP3 (NP_710161.1, 1 a.a. ∼ 236 a.a) full-length human protein. | |
| RUO | |
| 50511 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| synaptonemal complex protein 3 | |
| COR1/MGC71888/SCP3 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction