missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYF2 Antibody (OTI6A5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74413CL1
This item is not returnable.
View return policy
Description
SYF2 Monoclonal antibody specifically detects SYF2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| SYF2 | |
| Monoclonal | |
| CoraFluor 1 | |
| CBPIN, CCNDBP1 interactor, CCNDBP1-interactor, DKFZp564O2082, fSAP29, functional spliceosome-associated protein 29, GCIP-interacting protein p29, GCIPIP, NTC31, P29, pre-mRNA-splicing factor SYF2, SYF2 homolog, RNA splicing factor (S. cerevisiae) | |
| Human recombinant protein fragment corresponding to amino acids 2-92 of human SYF2(NP_056299) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 25949 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI6A5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction