Learn More
SYF2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16958-1-AP-20UL
 View more versions of this product
Product Code. 16816764
  • € 145.00 / 20µL
Description
This gene encodes a nuclear protein that interacts with cyclin D-type binding-protein 1, which is thought to be a cell cycle regulator at the G1/S transition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
SYF2
Polyclonal
Unconjugated
SYF2
CBPIN, CCNDBP1 interactor, fSAP29, GCIPIP, NTC31, P29, Pre mRNA splicing factor SYF2, SYF2
Rabbit
Antigen Affinity Chromatography
RUO
25949
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95926
SYF2
SYF2 Fusion Protein Ag10656