missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Symmetric Di-Methyl Arginine Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116813
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, MCF7, NIH/3T3, C6, Mouse liver, Rat liver.
Symmetrical dimethylarginine (SDMA) is the structural isomer of the endogenous nitric oxide synthase (NOS) inhibitor asymmetric dimethylarginine.
Specifications
| Symmetric Di-Methyl Arginine | |
| Polyclonal | |
| Unconjugated | |
| SDMA; Symmetric dimethylarginine | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| Dot Blot, ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Chemical, Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction