missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synapsin 3 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98603UV
This item is not returnable.
View return policy
Description
Synapsin 3 Polyclonal specifically detects Synapsin 3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Synapsin 3 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| synapsin IIIcN28H9.2 (synapsin III), synapsin-3 | |
| Produced in rabbits immunized with E. coli-derived Human Synapsin 3 fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 8224 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction