missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Synapsin II Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580087
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue.
Synapsins (synapsin I, II, III) are a family of synaptic vesicle phosphoproteins that have been implicated in modulation of neurotransmitter release and in synaptogenesis. Synapsin II (SYN2) is composed of two isoforms, synapsin IIa and IIb.
Specifications
| Synapsin II | |
| Polyclonal | |
| Unconjugated | |
| SYN2 | |
| 2900074L19Rik; AI836018; AI841723; SYN2; synapsin 2; synapsin II; Synapsin-2; SYNII; SYNIIa; SYNIIb | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20965, 29179, 6854 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| Q63537, Q64332, Q92777 | |
| SYN2 | |
| E. coli-derived human Synapsin II recombinant protein (Position: A497-D582). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction