missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Synapsin II Recombinant Rabbit Monoclonal Antibody (23GB4660), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | Synapsin II |
|---|---|
| Clone | 23GB4660 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089484
![]() |
Invitrogen™
MA552724 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087850
|
Invitrogen™
MA552725 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Synapsin II Recombinant Monoclonal antibody specifically detects Synapsin II in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| Synapsin II | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q92777 | |
| 6854 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4660 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2900074L19Rik; AI836018; AI841723; SYN2; synapsin 2; synapsin II; Synapsin-2; SYNII; SYNIIa; SYNIIb | |
| A synthesized peptide derived from human Synapsin II (500-582AA). | |
| Primary | |
| -20°C | |
| SYN2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title