missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptogyrin 1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06294MFV500
This item is not returnable.
View return policy
Description
Synaptogyrin 1 Polyclonal antibody specifically detects Synaptogyrin 1 in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Synaptogyrin 1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| MGC1939, synaptogyrin 1, synaptogyrin-1 | |
| Produced in rabbits immunized with E. coli-derived Human Synaptogyrin 1 fragment. | |
| 0.1 mL | |
| Neuroscience | |
| 9145 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction