missing translation for 'onlineSavingsMsg'
Learn More

Synaptogyrin 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 280.00 - € 540.75

Specifications

Antigen Synaptogyrin 1
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18499011
View Documents
Novus Biologicals
NBP1-87532-25ul
25 μL
€ 280.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18294587
View Documents
Novus Biologicals
NBP1-87532
0.1 mL
ONLINE EXCLUSIVE PRICE
€ 572.00
€ 540.75 / 0.10mL
Save € 31.25
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Synaptogyrin 1 Polyclonal specifically detects Synaptogyrin 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Synaptogyrin 1
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
9145
This antibody was developed against Recombinant Protein corresponding to amino acids:QRYQIGADSALFSQDYMDPSQDSSMPYAPYVEPTGPDPAGMGGTYQQPANTFDTEPQG
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Rabbit
Human
MGC1939, synaptogyrin 1, synaptogyrin-1
SYNGR1
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.