missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptogyrin 3 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77102C
This item is not returnable.
View return policy
Description
Synaptogyrin 3 Polyclonal antibody specifically detects Synaptogyrin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Synaptogyrin 3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| MGC20003, synaptogyrin 3, synaptogyrin-3 | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human SYNGR3. The immunogen is located within the last 50 amino acids of SYNGR3. Amino Acid Squence: ETYQSPFFTETLDTSPKGY | |
| 0.1 mL | |
| Stem Cell Markers | |
| 9143 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction