missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptojanin 2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22314AF350
This item is not returnable.
View return policy
Description
Synaptojanin 2 Polyclonal antibody specifically detects Synaptojanin 2 in Human samples. It is validated for Western Blot
Specifications
| Synaptojanin 2 | |
| Polyclonal | |
| Western Blot | |
| EC 3.1.3.36, inositol phosphate 5'-phosphatase 2, INPP5H, KIAA0348MGC44422, Synaptic inositol-14,5-trisphosphate 5-phosphatase 2, synaptojanin 2, synaptojanin-2 | |
| The immunogen this antibody was made to, maps to a region between residue 1275 to 1325 of human Synaptojanin 2 using the numbering given in entry NP_003889.1 (GeneID 8871). | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 8871 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction