missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Synaptojanin 2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15744691 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15744691 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15744691 Supplier Invitrogen™ Supplier No. PA556784

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: HGQYSILQTA RLLPGAPQQP PKARTGISKP YNVKQIKTTN AQEAEAAIRC LLEARGGASE EALSAVAPRD LEASSEP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

The gene is a member of the inositol-polyphosphate 5-phosphatase family. The encoded protein interacts with the ras-related C3 botulinum toxin substrate 1, which causes translocation of the encoded protein to the plasma membrane where it inhibits clathrin-mediated endocytosis. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptojanin 2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SYNJ2
Gene Accession No. O15056, O55207, Q9D2G5
Gene Alias AI481647; inositol phosphate 5'-phosphatase 2; inositol polyphosphate-5-phosphatase H; INPP5H; KIAA0348; LOW QUALITY PROTEIN: synaptojanin-2; mKIAA0348; phosphoinositide 5-phosphatase; SJ2; Synaptic inositol 1,4,5-trisphosphate 5-phosphatase 2; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 2; synaptojanin 2; synaptojanin-2; SYNJ2
Gene Symbols SYNJ2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Synaptojanin 2. Recombinant protein control fragment (Product #RP-94623).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20975, 84018, 8871
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.