missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptophysin Antibody (7H12), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47483C
This item is not returnable.
View return policy
Description
Synaptophysin Monoclonal specifically detects Synaptophysin in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Synaptophysin | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SYP | |
| Purified recombinant fragment of human Synaptophysin expressed in E. coli. [UniProt# P08247] | |
| 0.1 mL | |
| Angiogenesis, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers | |
| 6855 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| 7H12 | |
| Western Blot, ELISA, Immunohistochemistry-Paraffin | |
| Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction