missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptophysin Antibody (SYP/7976R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20528-20ug
This item is not returnable.
View return policy
Description
Synaptophysin Monoclonal antibody specifically detects Synaptophysin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Synaptophysin | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
| Recombinant fragment (around aa 274-313) of human Synaptophysin protein (exact sequence is proprietary) | |
| 20 μg | |
| Angiogenesis, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers | |
| 6855 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| SYP/7976R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction