missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Synaptophysin Recombinant Rabbit Monoclonal Antibody (RM258)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544590
This item is not returnable.
View return policy
Description
Synaptophysin Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.
Specifications
| Synaptophysin | |
| Recombinant Monoclonal | |
| 40 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| P08247 | |
| Syp | |
| A peptide corresponding to the C-terminus of Synaptophysin. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| RM258 | |
| Unconjugated | |
| Syp | |
| A230093K24Rik; AI848995; BM89 antigen; I79_012252; Major synaptic vesicle protein p38; MRX96; MRXSYP; p38; Syn; Synaptophysin; Syp; Syp I; Syp1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6855 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction