missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Synaptoporin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112735
This item is not returnable.
View return policy
Description
Synaptoporin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Synaptophysin 2, also known as synaptoporin, is a component of the synaptic vesicle membrane and thought to play an important role in synaptic vesicle trafficking (Dai et al., 2003). It is highly homologous to synaptophysin 1, yet differs in expression. Whereas synaptophysin 1 is ubiquitously expressed, synaptophysin 2 is concentrated in the mossy fiber synapses of the hippocampus (Grabs et al., 1994; Singec et al., 2002).
Specifications
| Synaptoporin | |
| Polyclonal | |
| Unconjugated | |
| SYNPR | |
| 1500003F20Rik; SPO; Synaptoporin; synaptorin; SYNPR | |
| Rabbit | |
| Protein G | |
| RUO | |
| 132204, 66030 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.35 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P22831, Q8TBG9 | |
| SYNPR | |
| Recombinant Human Synaptoporin protein (199-265AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction