missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptotagmin-11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech ProductsDescription
This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature.
Specifications
Specifications
| Antigen | Synaptotagmin-11 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SYT11 |
| Gene Accession No. | O08835, Q9BT88, Q9R0N3 |
| Gene Alias | KIAA0080, Synaptotagmin 11, synaptotagmin XI, Synaptotagmin-11, SYT11, SYT12, SytXI |
| Gene Symbols | Syt11 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?