missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNC Monoclonal Antibody (OTI5B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809588S
This item is not returnable.
View return policy
Description
SYNC Monoclonal antibody specifically detects SYNC in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SYNC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99742 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4861 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5B8 | |
| Unconjugated | |
| NPAS1 | |
| 1110057H03Rik; intermediate filament protein syncoilin; SNIP4; SYNC; SYNC1; synci; SYNCOILIN; syncoilin intermediate filament 1; syncoilin, intermediate filament 1; syncoilin, intermediate filament protein; syncoilin-1 | |
| Human recombinant protein fragment corresponding to amino acids 319-522 of human NPAS1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction