Learn More
Syndecan-1/CD138 Antibody (004), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90664MFV450
This item is not returnable.
View return policy
Description
Syndecan-1/CD138 Monoclonal antibody specifically detects Syndecan-1/CD138 in Mouse samples. It is validated for ELISA
Specifications
| ELISA | |
| Monoclonal | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Syndecan-1/CD138 (Accession#: NP_035649.1; Met1-Glu252). | |
| IgG |