Learn More
Syndecan-1/CD138 Antibody (004), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90664MFV450
Product Code. 18477949
  • € 663.00 / 0.10mL
Description
Syndecan-1/CD138 Monoclonal antibody specifically detects Syndecan-1/CD138 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA
Monoclonal
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Syndecan-1/CD138 (Accession#: NP_035649.1; Met1-Glu252).