missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syndecan-1/CD138 Antibody (359103), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2780CL1
This item is not returnable.
View return policy
Description
Syndecan-1/CD138 Monoclonal antibody specifically detects Syndecan-1/CD138 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunocytochemistry,CyTOF,Dual RNAscope ISH-IHC
Specifications
| Syndecan-1/CD138 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD138, CD138 antigen, SDC, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1 | |
| Mouse myeloma cell line NS0-derived recombinant human Syndecan-1/CD138, Gln18-Glu251, Accession # NP_002988 | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers | |
| 6382 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF, Dual RNAScope ISH-IHC | |
| 359103 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction