Learn More
Syndecan-1/CD138 Antibody (359103), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB2780MFV450
Product Code. 18575167
  • € 695.00 / 0.10mL
Description
Syndecan-1/CD138 Monoclonal antibody specifically detects Syndecan-1/CD138 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF, Dual RNAscope ISH-IHC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Syndecan-1/CD138
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Rat
0.1 mL
B Cell Development and Differentiation Markers
6382
Store at 4C in the dark.
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF, Dual RNAScope ISH-IHC
359103
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready, Dual RNAScope ISH-IHC
CD138, CD138 antigen, SDC, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1
Mouse myeloma cell line NS0-derived recombinant human Syndecan-1/CD138, Gln18-Glu251, Accession # NP_002988