missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syndecan-1/CD138 Antibody (B-A38), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14568AF594
This item is not returnable.
View return policy
Description
Syndecan-1/CD138 Monoclonal antibody specifically detects Syndecan-1/CD138 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| Syndecan-1/CD138 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| CD138, CD138 antigen, SDC, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1 | |
| U266 cell line | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers | |
| 6382 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| B-A38 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction