missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syndecan-1/CD138 Antibody (V92-P3D9*E7), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50314UV
This item is not returnable.
View return policy
Description
Syndecan-1/CD138 Monoclonal specifically detects Syndecan-1/CD138 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Syndecan-1/CD138 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SDC1 | |
| Peptide Sequence - PAGPSQADLHTP | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers | |
| 6382 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| V92-P3D9*E7 | |
| Western Blot, ELISA, Immunohistochemistry | |
| CD138, CD138 antigen, SDC, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction