Learn More
Syndecan-4 Antibody (047), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90700MFV500
Product Code. 18439039
  • € 674.00 / 0.10mL
Description
Syndecan-4 Monoclonal antibody specifically detects Syndecan-4 in Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Syndecan-4
Monoclonal
50mM Sodium Borate
Store at 4C in the dark.
IgG
Immunohistochemistry, Immunocytochemistry
047
6385