Learn More
Syndecan-4 Antibody (047), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90700MFV500
This item is not returnable.
View return policy
Description
Syndecan-4 Monoclonal antibody specifically detects Syndecan-4 in Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Syndecan-4 | |
| Monoclonal | |
| 50mM Sodium Borate | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunocytochemistry | |
| 047 | |
| 6385 | |
| Purified |