missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syndecan-4 Antibody (336304) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB29181JF525
This item is not returnable.
View return policy
Description
Syndecan-4 Monoclonal antibody specifically detects Syndecan-4 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Syndecan-4 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| amphiglycan, MGC22217, ryudocan, Ryudocan core protein, SYND4ryudocan amphiglycan, syndecan 4, syndecan 4 (amphiglycan, ryudocan), syndecan proteoglycan 4, syndecan-4 | |
| Mouse myeloma cell line NS0-derived recombinant human Syndecan-4, Ser20-Glu145, Accession # P31431 | |
| 0.1 mL | |
| Cellular Signaling, Neuroscience | |
| 6385 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 336304 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction