missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SynDIG4/PRRT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17261-1-AP-150UL
This item is not returnable.
View return policy
Description
PRRT1 is a protein coding gene.Specifications
| SynDIG4/PRRT1 | |
| Polyclonal | |
| Unconjugated | |
| PRRT1 | |
| C6orf31, NG5, PRRT1, SynDIG4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 260297, 406167, 80863 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35449, Q6MG82, Q99946 | |
| Prrt1 | |
| SynDIG4/PRRT1 Fusion Protein Ag10958 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction