missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98081-100ul
This item is not returnable.
View return policy
Description
SYNE1 Polyclonal antibody specifically detects SYNE1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SYNE1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| ARCA1, C6orf98, CPG2, dJ45H2.2, EC 4.2.1.49, EC 4.3.1.3, EDMD4, Enaptin, KIAA0796chromosome 6 open reading frame 98, KIAA1262FLJ41140, KIAA17568B, Myne-1, MYNE1DKFZp781J13156, Myocyte nuclear envelope protein 1, nesprin 1, nesprin-1, Nuclear envelope spectrin repeat protein 1, SCAR8FLJ30878, spectrin repeat containing, nuclear envelope 1, Synaptic nuclear envelope protein 1, synaptic nuclei expressed gene 1, Syne-1, SYNE-1B | |
| Produced in rabbits immunized with E. coli-derived Human SYNE1 fragment. | |
| 100 μg | |
| Developmental Biology | |
| 23345 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction