missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNE1 Monoclonal Antibody (OTI9A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811709
This item is not returnable.
View return policy
Description
SYNE1 gene encodes a spectrin repeat containing protein expressed in skeletal and smooth muscle, and peripheral blood lymphocytes, that localizes to the nuclear membrane. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia 8, also referred to as autosomal recessive cerebellar ataxia type 1 or recessive ataxia of Beauce. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| SYNE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NF91 | |
| SYNE1 | |
| Human recombit protein fragment corresponding to amino acids 1-285 of human SYNE1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI9A11 | |
| Unconjugated | |
| SYNE1 | |
| 8B, ARCA1, C6orf98, CPG2, EDMD4, KASH1, MYNE1, Nesp1, SCAR8, dJ45H2.2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23345 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction