Learn More
Invitrogen™ SYNE3 Recombinant Rabbit Monoclonal Antibody (23GB4670), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553785
 View more versions of this product
Product Code. 30089606
  • € 217.00 / 20µL
Description
As a component of the LINC (LInker of Nucleoskeleton and Cytoskeleton) complex involved in the connection between the nuclear lamina and the cytoskeleton. The nucleocytoplasmic interactions established by the LINC complex play an important role in the transmission of mechanical forces across the nuclear envelope and in nuclear movement and positioning. Probable anchoring protein which tethers the nucleus to the cytoskeleton by binding PLEC which can associate with the intermediate filament system. Plays a role in the regulation of aortic epithelial cell morphology, and is required for flow-induced centrosome polarization and directional migration in aortic endothelial cells. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SYNE3
Recombinant Monoclonal
Unconjugated
SYNE3
C14orf139; C14orf49; KASH domain-containing protein 3; KASH3; LINC00341; Nesp3; nesprin-3; NET53; Nuclear envelope spectrin repeat protein 3; RGD1304563; spectrin repeat containing nuclear envelope family member 3; spectrin repeat containing, nuclear envelope family member 3; SYNE3
Rabbit
Affinity chromatography
RUO
161176
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB4670
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6ZMZ3
SYNE3
A synthesized peptide derived from human Nesprin3 (750-935AA).