missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNJ2BP/ARIP2 Antibody (OTI3A9), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74417UV
This item is not returnable.
View return policy
Description
SYNJ2BP/ARIP2 Monoclonal specifically detects SYNJ2BP/ARIP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| SYNJ2BP/ARIP2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SYNJ2BP | |
| Full length recombinant protein of human SYNJ2BP/ARIP2 (NP_060843) produced in HEK293T cells. | |
| 0.1 mL | |
| 55333.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI3A9 | |
| Western Blot, Flow Cytometry | |
| activin receptor interacting protein 5, ARIP2, FLJ11271, Mitochondrial outer membrane protein 25, OMP25FLJ41973, synaptojanin 2 binding protein, synaptojanin-2-binding protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction