missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNJ2BP/ARIP2 Antibody (OTI3A9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01601
This item is not returnable.
View return policy
Description
SYNJ2BP/ARIP2 Monoclonal specifically detects SYNJ2BP/ARIP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| SYNJ2BP/ARIP2 | |
| Monoclonal | |
| 0.67 mg/mL | |
| Western Blot 1:1000-2000, Flow Cytometry 1:100 | |
| SYNJ2BP | |
| Full length human recombinant protein of human SYNJ2BP(NP_060843) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 55333 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry | |
| OTI3A9 | |
| Unconjugated | |
| activin receptor interacting protein 5, ARIP2, FLJ11271, Mitochondrial outer membrane protein 25, OMP25FLJ41973, synaptojanin 2 binding protein, synaptojanin-2-binding protein | |
| Mouse | |
| 15.7 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction