missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synphilin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 589.00
Specifications
| Antigen | Synphilin-1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18235391
|
Novus Biologicals
NBP2-55955 |
100 μL |
€ 589.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693628
|
Novus Biologicals
NBP2-55955-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Synphilin-1 Polyclonal specifically detects Synphilin-1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Synphilin-1 | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Neuroscience | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 9627 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RKASAVIHDQHKLSTEETEISPPLVKCGSAYEPENQSKDFLNKTFSDPHGRKVEKTTPDCQLRAFHLQSSAAESKPEEQVSGLNRTSSQGPEERSEYLK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Alpha-synuclein-interacting protein, MGC39814, Sph1, synphilin-1, synuclein, alpha interacting protein, SYPH1 | |
| SNCAIP | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title